Assay Details
| Gene Symbol: Ube3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191837.2 | NP_001178766.1 | 8 - 9 | 2608 | 131 | |
| XM_006229310.2 | XP_006229372.1 | 7 - 8 | 3453 | 131 | ||
| XM_006229313.2 | XP_006229375.1 | 6 - 7 | 3333 | 131 | ||
| XM_008759399.1 | XP_008757621.1 | 5 - 6 | 4163 | 131 | ||
| XM_008759400.1 | XP_008757622.1 | 7 - 8 | 2094 | 131 | ||
| XM_008759401.1 | XP_008757623.1 | 7 - 8 | 3397 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube3a
Genomic Map