Assay Details
| Gene Symbol: Siglech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759473.1 | XP_008757695.1 | 933 | 99 | ||
| GenBank mRNA | FQ232259.1 | - | 933 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Siglech| Gene Symbol: Siglech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759473.1 | XP_008757695.1 | 933 | 99 | ||
| GenBank mRNA | FQ232259.1 | - | 933 | 99 | ||
Set Membership: |
|