Assay Details
| Gene Symbol: Jazf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001065610.5 | XP_001065610.1 | 2 - 3 | 362 | 100 | |
| XM_006236535.2 | XP_006236597.1 | 2 - 3 | 1150 | 100 | ||
| XM_006236536.2 | XP_006236598.1 | 2 - 3 | 2268 | 100 | ||
| XM_008762953.1 | XP_008761175.1 | 2 - 3 | 489 | 100 | ||
| XM_008762954.1 | XP_008761176.1 | 3 - 4 | 704 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006236536.2, XM_008762953.1, XM_008762954.1 transcripts.
Set Membership: |
|

Jazf1
Genomic Map