Assay Details
Gene Symbol: Repin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236423.2 | XP_006236485.1 | 2 | 3584 | 73 | |
XM_006236424.2 | XP_006236486.1 | 2 | 3581 | 73 | ||
XM_006236425.2 | XP_006236487.1 | 3 | 3636 | 73 | ||
XM_006236426.2 | XP_006236488.1 | 3 | 3634 | 73 | ||
XM_006236427.2 | XP_006236489.1 | 3 | 3705 | 73 | ||
XM_006236428.2 | XP_006236490.1 | 2 | 3508 | 73 | ||
XM_006236429.2 | XP_006236491.1 | 2 | 3506 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
