Assay Details
| Gene Symbol: Fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013210.1 | NP_001013228.1 | 1 - 2 | 309 | 89 | |
| GenBank mRNA | BC083746.1 | - | 1 - 2 | 309 | 7192246 | 89 |
| FQ216652.1 | - | 1 - 2 | 356 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Fkbp14
Genomic Map