Assay Details
| Gene Symbol: Hoxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129878.1 | NP_001123350.1 | 1 - 2 | 757 | 90 | |
| GenBank mRNA | BC166717.1 | - | 1 - 2 | 752 | 7932029 | 90 |
| Gene Symbol: LOC100909443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749752.3 | XP_003749800.1 | 920 | 90 | ||
| Gene Symbol: NEWGENE_1564605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762951.1 | XP_008761173.1 | 1 - 2 | 1627 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Hoxa11
Genomic Map