Assay Details
Gene Symbol: Cycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012839.2 | NP_036971.1 | 2 - 3 | 248 | 151 | |
GenBank mRNA | BC081849.1 | - | 2 - 3 | 248 | 7192255 | 151 |
BC089051.1 | - | 2 - 3 | 259 | 7305562 | 151 | |
FQ211631.1 | - | 2 - 3 | 240 | 151 | ||
FQ212556.1 | - | 2 - 3 | 242 | 151 | ||
FQ212860.1 | - | 2 - 3 | 300 | 151 | ||
FQ213464.1 | - | 2 - 3 | 240 | 151 | ||
FQ214646.1 | - | 2 - 3 | 151 | |||
FQ214733.1 | - | 2 - 3 | 240 | 151 | ||
FQ214738.1 | - | 2 - 3 | 238 | 151 | ||
FQ214838.1 | - | 2 - 3 | 240 | 150 | ||
FQ214855.1 | - | 2 - 3 | 271 | 151 | ||
FQ214908.1 | - | 2 - 3 | 240 | 151 | ||
FQ215052.1 | - | 2 - 3 | 240 | 151 | ||
FQ215109.1 | - | 2 - 3 | 240 | 151 | ||
FQ215466.1 | - | 2 - 3 | 240 | 151 | ||
FQ216163.1 | - | 2 - 3 | 239 | 151 | ||
FQ216217.1 | - | 2 - 3 | 238 | 151 | ||
FQ216281.1 | - | 2 - 3 | 298 | 151 | ||
FQ216426.1 | - | 2 - 3 | 240 | 151 | ||
FQ216512.1 | - | 2 - 3 | 240 | 151 | ||
FQ216560.1 | - | 2 - 3 | 240 | 151 | ||
FQ216679.1 | - | 2 - 3 | 240 | 151 | ||
FQ217283.1 | - | 2 - 3 | 240 | 151 | ||
FQ217634.1 | - | 2 - 3 | 240 | 151 | ||
FQ218428.1 | - | 2 - 3 | 240 | 151 | ||
FQ218668.1 | - | 2 - 3 | 241 | 151 | ||
FQ220222.1 | - | 2 - 3 | 271 | 151 | ||
FQ220855.1 | - | 2 - 3 | 283 | 151 | ||
FQ223761.1 | - | 2 - 3 | 240 | 151 | ||
FQ224710.1 | - | 2 - 3 | 240 | 151 | ||
FQ224914.1 | - | 2 - 3 | 283 | 151 | ||
FQ224919.1 | - | 2 - 3 | 240 | 151 | ||
M20622.1 | - | 1 - 2 | 173 | 151 |
Gene Symbol: LOC100363502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751799.3 | XP_003751847.1 | 1 | 244 | 151 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
