Assay Details
Gene Symbol: Mum1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257301.2 | XP_006257363.1 | 3 - 4 | 482 | 84 | |
XM_006257302.2 | XP_006257364.1 | 4 - 5 | 432 | 84 | ||
XM_006257303.2 | XP_006257365.1 | 3 - 4 | 762 | 84 | ||
XM_006257304.2 | XP_006257366.1 | 2 - 3 | 705 | 84 | ||
GenBank mRNA | BC168654.1 | - | 1 - 2 | 272 | 7105971 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC168654.1, XM_006257301.2, XM_006257302.2, XM_006257303.2, XM_006257304.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
