Assay Details
| Gene Symbol: RGD1311558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760532.1 | XP_008758754.1 | 11 - 12 | 1186 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311558| Gene Symbol: RGD1311558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760532.1 | XP_008758754.1 | 11 - 12 | 1186 | 76 | |
Set Membership: |
|