Assay Details
Gene Symbol: Arl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022700.2 | NP_073191.1 | 2 - 3 | 276 | 120 | |
XM_006231559.2 | XP_006231621.1 | 3 - 4 | 797 | 120 | ||
GenBank mRNA | BC084722.1 | - | 2 - 3 | 276 | 7310623 | 120 |
U12568.1 | - | 2 - 3 | 280 | 120 | ||
X76921.1 | - | 3 - 4 | 408 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
