Assay Details
| Gene Symbol: Casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022260.3 | NP_071596.1 | 1 - 2 | 110 | 154 | |
| XM_008760546.1 | XP_008758768.1 | 2 - 3 | 703 | 154 | ||
| GenBank mRNA | AF072124.1 | - | 1 - 2 | 111 | 154 | |
| BC070936.1 | - | 1 - 2 | 111 | 7099111 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casp7
Genomic Map