Assay Details
Gene Symbol: Ccnj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106369.1 | NP_001099839.1 | 271 | 140 | ||
XM_006231388.2 | XP_006231450.1 | 306 | 140 | |||
XM_006231389.2 | XP_006231451.1 | 306 | 140 |
Gene Symbol: LOC100909771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749114.3 | XP_003749162.1 | 2 - 3 | 306 | 140 | |
XM_006231376.2 | XP_006231438.1 | 2 - 3 | 306 | 140 | ||
XM_006231377.2 | XP_006231439.1 | 2 - 3 | 306 | 140 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
