Assay Details
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011943.1 | NP_001011943.1 | 3 - 4 | 447 | 89 | |
XM_006231472.1 | XP_006231534.1 | 3 - 4 | 558 | 89 | ||
XM_006231474.2 | XP_006231536.1 | 3 - 4 | 559 | 89 | ||
GenBank mRNA | BC079096.1 | - | 3 - 4 | 447 | 7124109 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
