Assay Details
| Gene Symbol: Obfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011943.1 | NP_001011943.1 | 4 - 5 | 517 | 73 | |
| XM_006231472.1 | XP_006231534.1 | 4 - 5 | 628 | 73 | ||
| XM_006231474.2 | XP_006231536.1 | 4 - 5 | 629 | 73 | ||
| XM_006231475.2 | XP_006231537.1 | 2 - 3 | 2960 | 73 | ||
| GenBank mRNA | BC079096.1 | - | 4 - 5 | 517 | 7124109 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Obfc1
Genomic Map