Assay Details
Gene Symbol: Pprc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106363.1 | NP_001099833.1 | 6 - 7 | 3536 | 102 | |
XM_006231451.2 | XP_006231513.1 | 6 - 7 | 3557 | 102 | ||
XM_006231452.2 | XP_006231514.1 | 6 - 7 | 3556 | 102 | ||
XM_006231453.2 | XP_006231515.1 | 6 - 7 | 3553 | 102 | ||
XM_006231454.2 | XP_006231516.1 | 6 - 7 | 3557 | 102 | ||
XM_006231455.2 | XP_006231517.1 | 6 - 7 | 3548 | 102 | ||
XM_006231456.2 | XP_006231518.1 | 6 - 7 | 3472 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
