Assay Details
| Gene Symbol: Sfxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108527.1 | NP_001101997.1 | 12 - 13 | 847 | 63 | |
| XM_006231684.2 | XP_006231746.1 | 11 - 12 | 2908 | 63 | ||
| XM_006231685.2 | XP_006231747.1 | 11 - 12 | 1744 | 63 | ||
| XM_008760590.1 | XP_008758812.1 | 10 - 11 | 2642 | 63 | ||
| GenBank mRNA | FQ215895.1 | - | 12 - 13 | 836 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sfxn4
Genomic Map