Assay Details
| Gene Symbol: Cyp2c13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138514.1 | NP_612523.1 | 2 - 3 | 340 | 137 | |
| GenBank mRNA | BC088105.1 | - | 2 - 3 | 362 | 7372890 | 137 |
| BC098903.1 | - | 2 - 3 | 341 | 7373446 | 137 | |
| J02861.1 | - | 2 - 3 | 342 | 137 | ||
| M32277.1 | - | 2 - 3 | 340 | 137 | ||
| M33994.1 | - | 2 - 3 | 329 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2c13
Genomic Map