Assay Details
| Gene Symbol: Cacul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014248.1 | NP_001014270.1 | 3 - 4 | 876 | 95 | |
| XM_006231678.2 | XP_006231740.1 | 3 - 4 | 1036 | 95 | ||
| GenBank mRNA | BC083839.1 | - | 3 - 4 | 876 | 7114271 | 95 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P222475 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P453543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56306 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P222475 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P453543 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P453543 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P222475 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P222475 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P453543 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P222475 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P453543 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88884 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13635 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81998 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1594 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10731159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731163 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731166 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731160 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731157 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731158 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731164 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731165 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731167 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10731156 | - | - |
| Rat Genome 230 2.0 Array | 1374067_at | - | - |
| Rat Genome 230 2.0 Array | 1374067_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374067_at | - | - |
| Affymetrix HT Rat Focus Array | 1374067_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100064314.rn.1 | TC0100007987.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100064317.rn.1 | TC0100007987.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100064318.rn.1 | TC0100007987.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100064316.rn.1 | TC0100007987.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374067_at | - | - |
| Rat Expression Set 230 | 1374067_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10731156 | - | - |
| Rat Genome U34 Set | rc_AA852011_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22171221 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374308 | - | - |

Cacul1
Genomic Map