Assay Details
| Gene Symbol: Ahctf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271270.1 | NP_001258199.1 | 31 - 32 | 4690 | 81 | |
| XM_006250386.2 | XP_006250448.1 | 31 - 32 | 4737 | 81 | ||
| XM_008769858.1 | XP_008768080.1 | 31 - 32 | 4393 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ahctf1
Genomic Map