Assay Details
| Gene Symbol: Pycr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012208.1 | NP_001012208.1 | 3 - 4 | 436 | 72 | |
| GenBank mRNA | BC079222.1 | - | 3 - 4 | 436 | 7132148 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Pycr2
Genomic Map