Assay Details
| Gene Symbol: LOC317456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047894.1 | NP_001041359.1 | 2 | 546 | 61 | |
| GenBank mRNA | AY383668.1 | - | 2 | 546 | 61 | |
| Gene Symbol: Lpgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109376.2 | NP_001102846.1 | 5 - 6 | 945 | 61 | |
| XM_008769875.1 | XP_008768097.1 | 6 - 7 | 1003 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P032986 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P056970 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P236071 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P19819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22409 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P19819 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P056970 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P032986 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P236071 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P032986 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P056970 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P19819 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P236071 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P19819 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P236071 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P032986 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P056970 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P182671 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P236071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99494 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73623 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37088 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67965 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77900 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10766762 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933736 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933735 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933735 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933734 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766764 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766763 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10766760 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10933733 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10933733 | - | - |
| Rat Genome 230 2.0 Array | 1381193_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397880_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397880_at | - | - |
| Rat Genome 230 2.0 Array | 1389219_at | - | - |
| Rat Genome 230 2.0 Array | 1389219_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389358_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381193_at | - | - |
| Rat Genome 230 2.0 Array | 1389358_at | - | - |
| Affymetrix HT Rat Focus Array | 1389358_at | - | - |
| Affymetrix HT Rat Focus Array | 1397880_at | - | - |
| Affymetrix HT Rat Focus Array | 1381193_at | - | - |
| Affymetrix HT Rat Focus Array | 1389358_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397880_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381193_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300007543.rn.1 | TC1300001062.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007541.rn.1 | TC1300001062.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300007544.rn.1 | TC1300001062.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389358_at | - | - |
| Rat Expression Set 230 | 1389219_at | - | - |
| Rat Expression Set 230 | 1389219_PM_at | - | - |
| Rat Expression Set 230 | 1389358_PM_at | - | - |
| Rat Expression Set 230 | 1397880_at | - | - |
| Rat Expression Set 230 | 1381193_PM_at | - | - |
| Rat Expression Set 230 | 1381193_at | - | - |
| Rat Expression Set 230 | 1397880_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933733 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766760 | - | - |
| Rat Genome U34 Set | rc_AI030555_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21218126 | - | - |

LOC317456
Genomic Map