Assay Details
| Gene Symbol: Spata17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250449.2 | XP_006250511.1 | 6 - 7 | 638 | 66 | |
| XM_008769889.1 | XP_008768111.1 | 6 - 7 | 638 | 66 | ||
More Information
Set Membership: |
|

Spata17
Genomic Map