Assay Details
| Gene Symbol: Cr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005265.1 | NP_001005265.1 | 1 - 2 | 358 | 161 | |
| NM_001005330.1 | NP_001005330.1 | 358 | 161 | |||
| NM_019301.2 | NP_062174.1 | 1 - 2 | 358 | 161 | ||
| GenBank mRNA | BC061736.1 | - | 1 - 2 | 127 | 5599318 | 161 |
| D42114.1 | - | 1 - 2 | 167 | 161 | ||
| D42115.1 | - | 1 - 2 | 103 | 161 | ||
| L36532.1 | - | 1 - 2 | 126 | 161 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cr1l
Genomic Map