Assay Details
| Gene Symbol: H3f3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC086580.1 | - | 73 | 7307209 | 72 | |
| BC087725.1 | - | 43 | 7320825 | 72 | ||
| BC158615.1 | - | 59 | 7386252 | 72 | ||
| BC159439.1 | - | 75 | 9024967 | 72 | ||
| Gene Symbol: LOC100361558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728043.3 | XP_002728089.1 | 1 - 2 | 111 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086580.1,BC087725.1,BC158615.1,BC159439.1,XM_002728043.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

H3f3b
Genomic Map