Assay Details
| Gene Symbol: Traf3ip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014132.1 | NP_001014154.1 | 2 - 3 | 436 | 112 | |
| XM_008769865.1 | XP_008768087.1 | 1 - 2 | 1101 | 112 | ||
| XM_008769866.1 | XP_008768088.1 | 1 - 2 | 1101 | 112 | ||
| GenBank mRNA | BC089843.1 | - | 2 - 3 | 436 | 7368358 | 112 |
More Information
Updated Mapping Information
Set Membership: |
|

Traf3ip3
Genomic Map