Assay Details
| Gene Symbol: Dedd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250269.2 | XP_006250331.1 | 2 - 3 | 52 | 122 | |
| XM_006250272.2 | XP_006250334.1 | 2 - 3 | 132 | 122 | ||
| XM_006250273.2 | XP_006250335.1 | 1 - 2 | 58 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006250269.2,XM_006250272.2,XM_006250273.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dedd
Genomic Map