Assay Details
| Gene Symbol: Dedd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031800.1 | NP_113988.1 | 1 - 2 | 375 | 127 | |
| XM_006250269.2 | XP_006250331.1 | 3 - 4 | 440 | 127 | ||
| XM_006250270.2 | XP_006250332.1 | 1 - 2 | 390 | 127 | ||
| XM_006250272.2 | XP_006250334.1 | 3 - 4 | 520 | 127 | ||
| XM_006250273.2 | XP_006250335.1 | 2 - 3 | 446 | 127 | ||
| XM_006250274.2 | XP_006250336.1 | 1 - 2 | 394 | 127 | ||
| GenBank mRNA | AF053362.1 | - | 1 - 2 | 375 | 127 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dedd
Genomic Map