Assay Details
Gene Symbol: Ogfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053340.2 | NP_445792.2 | 3 - 4 | 325 | 138 | |
GenBank mRNA | BC101898.1 | - | 3 - 4 | 306 | 7932932 | 138 |
EV778659.1 | - | 3 - 4 | 325 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
