Assay Details
| Gene Symbol: Phactr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214459.2 | NP_999624.1 | 10 - 11 | 1477 | 87 | |
| GenBank mRNA | AY500157.1 | - | 10 - 11 | 1477 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40708 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017500 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017500 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017500 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20723 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67410 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93847 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24995 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10842696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842700 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842701 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10842695 | - | - |
| Rat Genome 230 2.0 Array | 1380054_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393307_at | - | - |
| Rat Genome 230 2.0 Array | 1393307_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380054_at | - | - |
| Affymetrix HT Rat Focus Array | 1380054_at | - | - |
| Affymetrix HT Rat Focus Array | 1380054_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300018634.rn.1 | TC0300002532.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393307_at | - | - |
| Rat Expression Set 230 | 1393307_PM_at | - | - |
| Rat Expression Set 230 | 1380054_PM_at | - | - |
| Rat Expression Set 230 | 1380054_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10842695 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376553 | - | - |

Phactr3
Genomic Map