Assay Details
| Gene Symbol: Cyp3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153312.2 | NP_695224.2 | 138 | 76 | ||
| GenBank mRNA | BC089765.1 | - | 143 | 7372112 | 76 | |
| M13646.1 | - | 138 | 76 | |||
| U09742.1 | - | 140 | 76 | |||
| X79320.1 | - | 140 | 76 | |||
More Information
Set Membership: |
|

Cyp3a2
Genomic Map