Assay Details
| Gene Symbol: Cyp3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153312.2 | NP_695224.2 | 868 | 157 | ||
| GenBank mRNA | BC089765.1 | - | 873 | 7372112 | 157 | |
| M13646.1 | - | 868 | 157 | |||
| U09742.1 | - | 870 | 157 | |||
| X79320.1 | - | 870 | 157 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Cyp3a2
Genomic Map