Assay Details
| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044297.1 | NP_001037762.1 | 1 - 2 | 254 | 60 | |
| GenBank mRNA | BC082099.1 | - | 1 - 2 | 254 | 7134466 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd61
Genomic Map