Assay Details
| Gene Symbol: Exoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024770.1 | NP_001019941.1 | 13 - 14 | 1773 | 98 | |
| GenBank mRNA | BC097392.1 | - | 13 - 14 | 1773 | 7453203 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P414757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29542 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P414757 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P414757 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P414757 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P414757 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77356 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18145 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76843 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10776405 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776400 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776411 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776413 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776403 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776407 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776401 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776410 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776414 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776412 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776406 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776408 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776404 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776402 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10776395 | - | - |
| Rat Genome 230 2.0 Array | 1375749_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383387_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393905_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383386_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1393016_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1383388_at | - | - |
| Rat Genome 230 2.0 Array | 1383388_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383387_at | - | - |
| Rat Genome 230 2.0 Array | 1393016_s_at | - | - |
| Rat Genome 230 2.0 Array | 1393905_at | - | - |
| Rat Genome 230 2.0 Array | 1383386_a_at | - | - |
| Rat Genome 230 2.0 Array | 1375749_at | - | - |
| Affymetrix HT Rat Focus Array | 1383386_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1375749_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383387_at | - | - |
| Affymetrix HT Rat Focus Array | 1383388_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383386_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1383388_at | - | - |
| Affymetrix HT Rat Focus Array | 1375749_at | - | - |
| Affymetrix HT Rat Focus Array | 1383387_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400012132.rn.1 | TC1400001645.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400012134.rn.1 | TC1400001645.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400012131.rn.1 | TC1400001645.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400012135.rn.1 | TC1400001645.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375749_PM_at | - | - |
| Rat Expression Set 230 | 1375749_at | - | - |
| Rat Expression Set 230 | 1393016_s_at | - | - |
| Rat Expression Set 230 | 1393905_at | - | - |
| Rat Expression Set 230 | 1383386_PM_a_at | - | - |
| Rat Expression Set 230 | 1383387_at | - | - |
| Rat Expression Set 230 | 1393016_PM_s_at | - | - |
| Rat Expression Set 230 | 1393905_PM_at | - | - |
| Rat Expression Set 230 | 1383386_a_at | - | - |
| Rat Expression Set 230 | 1383387_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10776395 | - | - |
| Rat Genome U34 Set | rc_AI145446_at | - | - |
| Rat Genome U34 Set | rc_AI177465_at | - | - |
| Rat Genome U34 Set | H33610_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368953 | - | - |

Exoc1
Genomic Map