Assay Details
| Gene Symbol: Noa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006959.1 | NP_001006960.1 | 1 - 2 | 1226 | 122 | |
| XM_006250845.2 | XP_006250907.1 | 1 - 2 | 1541 | 122 | ||
| GenBank mRNA | BC083902.1 | - | 1 - 2 | 1226 | 7133605 | 122 |
More Information
Updated Mapping Information
Set Membership: |
|

Noa1
Genomic Map