Assay Details
| Gene Symbol: Cenpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004098.1 | NP_001004098.1 | 15 - 16 | 2352 | 109 | |
| GenBank mRNA | AY693786.1 | - | 8 - 9 | 1195 | 109 | |
| AY693788.1 | - | 15 - 16 | 2352 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpc
Genomic Map