Assay Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033652.1 | NP_001028824.1 | 1 - 2 | 249 | 128 | |
| GenBank mRNA | AF453776.1 | - | 1 - 2 | 249 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF453776.1, NM_001033652.1 transcripts.
Set Membership: |
|

Tmprss11d
Genomic Map