Assay Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033652.1 | NP_001028824.1 | 2 - 3 | 378 | 137 | |
| GenBank mRNA | AF453776.1 | - | 2 - 3 | 378 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11d
Genomic Map