Assay Details
Gene Symbol: Cyp2c23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031839.3 | NP_114027.2 | 6 - 7 | 992 | 108 | |
GenBank mRNA | BC078707.1 | - | 6 - 7 | 992 | 7110580 | 108 |
FQ209508.1 | - | 4 - 5 | 591 | 108 | ||
FQ218733.1 | - | 1 - 2 | 119 | 108 | ||
FQ218882.1 | - | 3 - 4 | 341 | 108 | ||
U04733.1 | - | 6 - 7 | 973 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
