Assay Details
Gene Symbol: Fam160b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_342068.5 | XP_342069.4 | 12 - 13 | 1706 | 67 | |
GenBank mRNA | FQ234191.1 | - | 1 - 2 | 104 | 67 | |
FQ234370.1 | - | 1 - 2 | 104 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
