Assay Details
Gene Symbol: Cxcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030845.1 | NP_110472.1 | 2 - 3 | 243 | 111 | |
GenBank mRNA | D11444.1 | - | 2 - 3 | 228 | 111 | |
M86536.1 | - | 2 - 3 | 243 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
