Assay Details
| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182952.2 | NP_891997.1 | 1 - 2 | 124 | 98 | |
| GenBank mRNA | AF537209.1 | - | 1 - 2 | 124 | 98 | |
| AY340181.1 | - | 1 - 2 | 103 | 98 | ||
| BC168725.1 | - | 1 - 2 | 132 | 6919932 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF537209.1, AY340181.1, BC168725.1, NM_182952.2 transcripts.
Set Membership: |
|

Cxcl11
Genomic Map