Assay Details
| Gene Symbol: Cxcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250721.2 | XP_006250783.1 | 3 - 4 | 509 | 99 | |
| GenBank mRNA | D87927.1 | - | 3 - 4 | 279 | 99 | |
More Information
Set Membership: |
|

Cxcl3
Genomic Map