Assay Details
| Gene Symbol: Cxcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138522.1 | NP_612531.1 | 4 - 5 | 415 | 129 | |
| GenBank mRNA | D21095.1 | - | 4 - 5 | 415 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl3
Genomic Map