Assay Details
| Gene Symbol: Apex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024148.1 | NP_077062.1 | 1 - 2 | 48 | 76 | |
| GenBank mRNA | BC078816.1 | - | 1 - 2 | 56 | 7127205 | 76 |
| D44495.1 | - | 1 - 2 | 48 | 76 | ||
| FQ220521.1 | - | 1 - 2 | 50 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC078816.1,D44495.1,NM_024148.1 transcripts.
This assay is in the 5'UTR region of the BC078816.1,D44495.1,NM_024148.1 transcripts.
Set Membership: |
|

Apex1
Genomic Map