Assay Details
| Gene Symbol: Mapk1ip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108373.1 | NP_001101843.1 | 2 - 3 | 143 | 102 | |
| XM_006251812.2 | XP_006251874.1 | 2 - 3 | 141 | 102 | ||
| GenBank mRNA | BC167004.1 | - | 2 - 3 | 141 | 9090872 | 102 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC167004.1, NM_001108373.1, XM_006251812.2 transcripts.
Set Membership: |
|

Mapk1ip1l
Genomic Map