Assay Details
| Gene Symbol: Slc39a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107260.1 | NP_001100730.1 | 2 - 3 | 368 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc39a2| Gene Symbol: Slc39a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107260.1 | NP_001100730.1 | 2 - 3 | 368 | 84 | |
Set Membership: |
|