Assay Details
Gene Symbol: Pnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106031.1 | NP_001099501.1 | 3 - 4 | 358 | 70 | |
XM_006251857.2 | XP_006251919.1 | 3 - 4 | 388 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
