Assay Details
| Gene Symbol: Samd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107254.1 | NP_001100724.1 | 10 - 11 | 2087 | 61 | |
| XM_008770515.1 | XP_008768737.1 | 10 - 11 | 2964 | 61 | ||
| XM_008770516.1 | XP_008768738.1 | 9 - 10 | 2699 | 61 | ||
| GenBank mRNA | BC168894.1 | - | 10 - 11 | 2108 | 7453870 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Samd4a
Genomic Map