Assay Details
| Gene Symbol: Tmem63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134496.2 | NP_001127968.2 | 9 - 10 | 840 | 61 | |
| XM_006250364.2 | XP_006250426.1 | 9 - 10 | 1017 | 61 | ||
| XM_006250365.2 | XP_006250427.1 | 9 - 10 | 872 | 61 | ||
| GenBank mRNA | FQ210846.1 | - | 9 - 10 | 812 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem63a
Genomic Map