Assay Details
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107199.1 | NP_001100669.1 | 17 - 18 | 2097 | 66 | |
GenBank mRNA | AY724501.1 | - | 17 - 18 | 2086 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P632328 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20220 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083185 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P632328 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P632328 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083185 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P632328 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083185 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P632328 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24789 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88718 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86391 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10770618 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770617 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770630 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770632 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770631 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770629 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770625 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770621 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770628 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770623 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770616 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770614 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770620 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770622 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770633 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770627 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770624 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770626 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770619 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770615 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10770613 | - | - |
Rat Genome 230 2.0 Array | 1373119_PM_at | - | - |
Rat Genome 230 2.0 Array | 1395941_PM_at | - | - |
Rat Genome 230 2.0 Array | 1395941_at | - | - |
Rat Genome 230 2.0 Array | 1373119_at | - | - |
Affymetrix HT Rat Focus Array | 1373119_at | - | - |
Affymetrix HT Rat Focus Array | 1373119_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395941_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395941_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300015477.rn.1 | TC1300002180.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300015478.rn.1 | TC1300002180.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300015479.rn.1 | TC1300002180.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300015480.rn.1 | TC1300002180.rn.1 | RTA TC |
Rat Expression Set 230 | 1373119_PM_at | - | - |
Rat Expression Set 230 | 1373119_at | - | - |
Rat Expression Set 230 | 1395941_PM_at | - | - |
Rat Expression Set 230 | 1395941_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10770613 | - | - |
Rat Genome U34 Set | rc_AI071931_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22160166 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361373 | - | - |
